Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026106.4 | NP_080382.2 | 1 | 866 | ||
GenBank mRNA | AK007230.1 | - | 1 | 841 | ||
AK031057.1 | - | 1 | 800 | |||
AK075713.1 | - | 1 | 866 | |||
AK090113.1 | - | 1 | 848 | |||
AK139866.1 | - | 1 | 796 | |||
AK144749.1 | - | 1 | 860 | |||
AK146573.1 | - | 1 | 829 | |||
AK178588.1 | - | 1 | 86 | |||
AK211344.1 | - | 1 | 86 | |||
BC013461.1 | - | 1 | 435 | 4222118 |