Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dsg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030596.3 | NP_085099.2 | 16 | 3260 | ||
GenBank mRNA | AK037142.1 | - | 16 | 3291 | ||
AK148657.1 | - | 16 | 3261 | |||
BC132058.1 | - | 16 | 3289 | 40130335 | ||
BC132639.1 | - | 16 | 3289 | 40130916 |