Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apobec1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134391.1 | NP_001127863.1 | 6 | 922 | ||
NM_031159.3 | NP_112436.1 | 6 | 978 | |||
GenBank mRNA | AK146427.1 | - | 6 | 854 | ||
AK151799.1 | - | 6 | 850 | |||
AK165209.1 | - | 6 | 861 | |||
AK166412.1 | - | 6 | 854 | |||
BC003792.1 | - | 6 | 866 | 3494195 | ||
U22262.1 | - | 6 | 978 | |||
U22263.1 | - | 6 | 922 | |||
U22264.1 | - | 3 | 678 |