Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111059.1 | NP_001104529.1 | 2 | 350 | ||
NM_133654.3 | NP_598415.1 | 2 | 350 | |||
GenBank mRNA | AK044121.1 | - | 2 | 286 | ||
AK051507.1 | - | 2 | 266 | |||
AK053874.1 | - | 2 | 286 | |||
AK089010.1 | - | 2 | 256 | |||
AK167329.1 | - | 2 | 261 | |||
AK167350.1 | - | 2 | 261 | |||
BC006607.1 | - | 2 | 266 | 3966337 | ||
BC066820.1 | - | 2 | 223 | |||
BY249301.1 | - | 2 | 353 | |||
BY327943.1 | - | 2 | 303 | |||
S69293.1 | - | 2 | 228 | |||
S69299.1 | - | 2 | 205 | |||
S69301.1 | - | 2 | 205 |