Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133655.2 | NP_598416.1 | 8 | 1299 | ||
GenBank mRNA | AK136370.1 | - | 8 | 1264 | ||
AK170741.1 | - | 8 | 1264 | |||
BC011433.1 | - | 8 | 1253 | 2811801 | ||
X59047.1 | - | 8 | 1018 |