Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cab39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133781.4 | NP_598542.3 | 9 | 2128 | ||
GenBank mRNA | AK005226.1 | - | 1 | 55 | ||
AK144665.1 | - | 10 | 1961 | |||
BC020041.1 | - | 9 | 1791 | 3990872 | ||
BC029053.1 | - | 5 | 1302 | |||
S51858.1 | - | 9 | 1866 |