Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144900.2 | NP_659149.1 | 6 | 806 | ||
GenBank mRNA | AK088000.1 | - | 6 | 767 | ||
AK137001.1 | - | 6 | 765 | |||
AK159279.1 | - | 6 | 763 | |||
AK159306.1 | - | 6 | 761 | |||
AK181532.1 | - | 2 | 140 | |||
AK203787.1 | - | 375 | ||||
AK206258.1 | - | 3 | 305 | |||
BC010319.1 | - | 4 | 391 | |||
BC021496.1 | - | 6 | 756 | 5346159 | ||
BC023794.1 | - | 7 | 701 | 5351943 | ||
BC023964.1 | - | 6 | 746 | |||
BC025618.1 | - | 6 | 765 | 5320976 | ||
BC025627.1 | - | 6 | 756 | 5320513 | ||
BC025811.1 | - | 6 | 743 | 5096872 | ||
BC032187.1 | - | 6 | 806 | 5321268 | ||
BC033435.1 | - | 6 | 702 | 4950746 | ||
BC033471.1 | - | 6 | 746 | 4954119 | ||
BC034177.1 | - | 5 | 511 | |||
BC035352.1 | - | 5 | 476 | |||
BC042435.1 | - | 6 | 756 | 4951288 |