Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153150.2 | NP_694790.1 | 8 | 904 | ||
GenBank mRNA | AK159289.1 | - | 8 | 867 | ||
AK170191.1 | - | 8 | 868 | |||
BC037087.1 | - | 8 | 864 | 5124403 |