Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005328.2 | NP_005319.1 | 4 | 2549 | ||
GenBank mRNA | U54804.1 | - | 4 | 2546 |