Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acadm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007382.5 | NP_031408.1 | 2 | 322 | ||
GenBank mRNA | AK008149.1 | - | 2 | 124 | ||
AK131793.1 | - | 2 | 124 | |||
AK146444.1 | - | 2 | 126 | |||
BC013498.1 | - | 2 | 107 | 4222450 | ||
CX232934.1 | - | 2 | 313 | |||
U07159.1 | - | 2 | 111 |