Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acads | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007383.3 | NP_031409.2 | 10 | 1374 | ||
GenBank mRNA | AK155361.1 | - | 10 | 1353 | ||
AK169428.1 | - | 10 | 1311 | |||
BC016259.1 | - | 10 | 1341 | 4913813 | ||
BC055063.1 | - | 9 | 1527 | |||
L11163.1 | - | 10 | 1298 |