Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acvr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110204.1 | NP_001103674.1 | 13 | 2162 | ||
NM_001110205.1 | NP_001103675.1 | 11 | 1640 | |||
NM_007394.3 | NP_031420.2 | 12 | 1917 | |||
GenBank mRNA | AF332087.1 | - | 10 | 1550 | ||
AF332088.1 | - | 10 | 1550 | |||
AK053428.1 | - | 13 | 2161 | |||
AK150014.1 | - | 12 | 1917 | |||
AK150075.1 | - | 12 | 1917 | |||
BC058718.1 | - | 10 | 1638 | 6418954 | ||
L15436.1 | - | 11 | 1990 |