Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMGB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301228.1 | NP_001288157.1 | 5 | 814 | ||
NM_001301229.1 | NP_001288158.1 | 5 | 705 | |||
NM_001301231.1 | NP_001288160.1 | 5 | 744 | |||
NM_005342.3 | NP_005333.2 | 5 | 709 | |||
GenBank mRNA | BC037210.1 | - | 4 | 614 | ||
BC070482.1 | - | 6 | 793 | 6140626 | ||
BX537505.1 | - | 3 | 2105 | |||
Y10043.1 | - | 5 | 688 |