Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007400.2 | NP_031426.2 | 23 | 3864 | ||
GenBank mRNA | AK051780.1 | - | 13 | 2678 | ||
AK220274.1 | - | 10 | 2074 | |||
D50411.1 | - | 23 | 3864 |