Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adrb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007420.3 | NP_031446.2 | 1 | 1540 | ||
GenBank mRNA | AK080241.1 | - | 1 | 1538 | ||
AK080276.1 | - | 1 | 1540 | |||
BC030346.1 | - | 1 | 976 | |||
BC032883.1 | - | 1 | 1518 | 1349283 |
Set Membership: |
G-protein coupled receptor |