Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005368.2 | NP_005359.1 | 2 | 268 | ||
NM_203377.1 | NP_976311.1 | 3 | 360 | |||
NM_203378.1 | NP_976312.1 | 3 | 343 | |||
XM_005261605.2 | XP_005261662.1 | 3 | 304 | |||
GenBank mRNA | AK300657.1 | - | 3 | 226 | ||
AK314057.1 | - | 3 | 329 | |||
BC014547.1 | - | 2 | 264 | 4244838 | ||
BC018001.1 | - | 2 | 233 | 4271145 | ||
BQ925917.1 | - | 392 | ||||
BQ956082.1 | - | 247 | ||||
BU585249.1 | - | 2 | 390 | |||
CB140824.1 | - | 3 | 373 | |||
CR456516.1 | - | 2 | 236 | |||
CR541949.1 | - | 2 | 188 |