Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ager | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271422.1 | NP_001258351.1 | 7 | 763 | ||
NM_001271423.1 | NP_001258352.1 | 7 | 736 | |||
NM_001271424.1 | NP_001258353.1 | 6 | 858 | |||
NM_007425.3 | NP_031451.2 | 7 | 763 | |||
GenBank mRNA | AB207883.1 | - | 7 | 726 | ||
AK004802.1 | - | 7 | 753 | |||
BC061182.1 | - | 7 | 751 | 30300427 | ||
EU520325.1 | - | 7 | 719 | |||
EU570240.1 | - | 7 | 719 | |||
EU570241.1 | - | 6 | 914 | |||
EU570242.1 | - | 7 | 719 | |||
EU570243.1 | - | 7 | 719 | |||
EU570244.1 | - | 6 | 914 | |||
EU570245.1 | - | 6 | 914 | |||
EU570246.1 | - | 6 | 876 | |||
EU570247.1 | - | 6 | 914 | |||
EU906863.1 | - | 7 | 692 | |||
EU906864.1 | - | 6 | 914 | |||
GU164727.1 | - | 6 | 833 | |||
GU164728.1 | - | 7 | 738 | |||
GU164730.1 | - | 7 | 738 | |||
GU164731.1 | - | 7 | 741 | |||
KC692918.1 | - | 7 | 719 | |||
L33412.1 | - | 7 | 727 |