Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh3a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001112725.1 | NP_001106196.1 | 7 | 929 | ||
NM_007436.2 | NP_031462.2 | 7 | 1061 | |||
GenBank mRNA | AF072815.1 | - | 7 | 967 | ||
AK144241.1 | - | 7 | 932 | |||
BC144946.1 | - | 7 | 961 | 9053494 | ||
BC150677.1 | - | 7 | 961 | 9087588 | ||
EL959139.1 | - | 2 | 208 | |||
EL963960.1 | - | 3 | 409 | |||
U12785.1 | - | 7 | 1058 |