Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldoa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177307.1 | NP_001170778.1 | 5 | 672 | ||
NM_001177308.1 | NP_001170779.1 | 4 | 545 | |||
NM_007438.4 | NP_031464.1 | 4 | 595 | |||
GenBank mRNA | AK002561.1 | - | 4 | 484 | ||
AK145639.1 | - | 4 | 482 | |||
AK154295.1 | - | 4 | 478 | |||
AK162290.1 | - | 4 | 481 | |||
BC043026.1 | - | 4 | 471 | 5691975 | ||
BC050896.1 | - | 4 | 513 | 5715436 | ||
BC066218.1 | - | 4 | 491 | 30054842 | ||
BC066801.1 | - | 4 | 491 | 30072521 | ||
BC089495.1 | - | 4 | 524 | 30312668 | ||
CF617557.1 | - | 4 | 431 | |||
EF662059.1 | - | 4 | 526 | |||
EF662060.1 | - | 4 | 476 | |||
X03797.1 | - | 2 | 70 | |||
Y00516.1 | - | 4 | 441 |