Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ikzf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK040268.1 | - | 5 | 628 | ||
AK077815.1 | - | 6 | 1192 | |||
AK154175.1 | - | 6 | 917 | |||
AK220235.1 | - | 6 | 861 | |||
AY536269.1 | - | 3 | 205 | |||
BC138789.1 | - | 5 | 426 | 8861811 | ||
L03547.1 | - | 5 | 411 | |||
S74708.1 | - | 3 | 472 |