Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001112703.2 | NP_001106174.1 | 2 | 1468 | ||
NM_001283045.1 | NP_001269974.1 | 2 | 345 | |||
NM_001283046.1 | NP_001269975.1 | 3 | 1673 | |||
NM_001283047.1 | NP_001269976.1 | 3 | 1612 | |||
NM_009594.4 | NP_033724.2 | 2 | 239 | |||
GenBank mRNA | AK088252.1 | - | 3 | 1598 | ||
AK090095.1 | - | 2 | 197 | |||
AK166028.1 | - | 2 | 1457 | |||
AK166210.1 | - | 3 | 1707 | |||
AK192334.1 | - | 298 | ||||
AK194745.1 | - | 2 | 176 | |||
BC059260.1 | - | 2 | 266 | 6408990 | ||
BC103770.1 | - | 2 | 692 | 30848553 | ||
BY152805.1 | - | 2 | 238 | |||
CJ099623.1 | - | 2 | 190 | |||
CJ101520.1 | - | 2 | 240 | |||
CJ128107.1 | - | 2 | 234 | |||
J02995.1 | - | 2 | 192 | |||
K03228.1 | - | 2 | 234 | |||
M12263.1 | - | 3 | 599 | |||
M12264.1 | - | 2 | 345 | |||
M12265.1 | - | 2 | 394 |