Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrnb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009601.4 | NP_033731.3 | 4 | 317 | ||
GenBank mRNA | AK087554.1 | - | 4 | 296 | ||
BC023318.1 | - | 4 | 323 | 4009493 | ||
BY771238.1 | - | 3 | 252 | |||
CJ176855.1 | - | 4 | 317 | |||
M14537.1 | - | 4 | 260 |