Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adipoq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009605.4 | NP_033735.3 | 3 | 821 | ||
GenBank mRNA | AK003138.1 | - | 3 | 739 | ||
AK041214.1 | - | 3 | 771 | |||
AK134112.1 | - | 3 | 821 | |||
AX767955.1 | - | 2 | 723 | |||
AX767957.1 | - | 3 | 752 | |||
AY749429.1 | - | 2 | 707 | |||
AY754346.1 | - | 2 | 707 | |||
BC028770.1 | - | 3 | 736 | 1329799 | ||
U37222.1 | - | 3 | 752 | |||
U49915.1 | - | 2 | 723 |