Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110778.1 | NP_001104248.1 | 26 | 3077 | ||
NM_009613.2 | NP_033743.2 | 27 | 2595 | |||
GenBank mRNA | AB009676.1 | - | 27 | 2428 | ||
BC054536.1 | - | 26 | 2937 | 6400084 | ||
CD353697.1 | - | 3 | 196 |