Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037722.2 | NP_001032811.2 | 7, 8 | 809 | ||
NM_009614.2 | NP_033744.1 | 7, 8 | 809 | |||
GenBank mRNA | AF006196.1 | - | 7, 8 | 770 | ||
AK039384.1 | - | 7, 8 | 789 | |||
AK048901.1 | - | 7, 8 | 796 | |||
AK142520.1 | - | 7, 8 | 791 | |||
AK149684.1 | - | 7, 8 | 790 | |||
AK149796.1 | - | 782 | ||||
AK151804.1 | - | 7, 8 | 745 | |||
AK152725.1 | - | 7, 8 | 745 | |||
AK152954.1 | - | 7, 8 | 746 | |||
AK152973.1 | - | 7, 8 | 746 | |||
AK170101.1 | - | 7, 8 | 775 | |||
AK170365.1 | - | 7, 8 | 789 | |||
AK179173.1 | - | 2, 3 | 172 | |||
AK205473.1 | - | 2, 3 | 172 | |||
BC009132.1 | - | 7, 8 | 758 | 3152945 | ||
BC057909.1 | - | 6, 7 | 951 | |||
EF506571.1 | - | 1, 2 | 70 |