Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037722.2 | NP_001032811.2 | 16 | 2099 | ||
NM_009614.2 | NP_033744.1 | 16 | 2099 | |||
GenBank mRNA | AF006196.1 | - | 16 | 2060 | ||
AK039384.1 | - | 16 | 2079 | |||
AK048901.1 | - | 16 | 2086 | |||
AK142520.1 | - | 16 | 2082 | |||
AK149684.1 | - | 16 | 2081 | |||
AK149796.1 | - | 2072 | ||||
AK151804.1 | - | 16 | 2035 | |||
AK152725.1 | - | 16 | 2035 | |||
AK152954.1 | - | 16 | 2038 | |||
AK152973.1 | - | 16 | 2037 | |||
AK170101.1 | - | 16 | 2065 | |||
AK170365.1 | - | 16 | 2079 | |||
AK206889.1 | - | 3 | 363 | |||
AK218324.1 | - | 2 | 215 | |||
BC057909.1 | - | 12 | 2650 | |||
EF506571.1 | - | 10 | 1360 |