Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291890.1 | NP_001278819.1 | 4 | 267 | ||
NM_001291891.1 | NP_001278820.1 | 4 | 452 | |||
NM_009616.4 | NP_033746.1 | 4 | 452 | |||
GenBank mRNA | AF019887.2 | - | 4 | 357 | ||
AK147217.1 | - | 4 | 375 | |||
AK147549.1 | - | 4 | 268 | |||
BC093480.1 | - | 4 | 443 | |||
D50410.2 | - | 4 | 384 |