Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PODXL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018111.2 | NP_001018121.1 | 9 | 5260 | ||
NM_005397.3 | NP_005388.2 | 8 | 5164 | |||
GenBank mRNA | BC042466.1 | - | 7 | 5452 | ||
BX641124.1 | - | 3 | 5449 | |||
U97519.1 | - | 8 | 5164 |