Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009618.2 | NP_033748.2 | 6 | 505 | ||
GenBank mRNA | AK016550.1 | - | 6 | 505 | ||
AK161261.1 | - | 6 | 470 | |||
AK161266.1 | - | 6 | 494 | |||
U16242.1 | - | 6 | 494 | |||
U22057.1 | - | 5 | 445 | |||
U38806.1 | - | 6 | 510 |