Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKAB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005399.4 | NP_005390.1 | 5 | 675 | ||
NR_103871.1 | 4 | 508 | ||||
XM_011509729.1 | XP_011508031.1 | 5 | 675 | |||
GenBank mRNA | AJ224538.1 | - | 4 | 491 | ||
AK292820.1 | - | 5 | 556 | |||
AK294863.1 | - | 4 | 480 | |||
AK310982.1 | - | 5 | 556 | |||
AK316005.1 | - | 4 | 386 | |||
AL037763.1 | - | 4 | 472 | |||
BC053610.1 | - | 5 | 630 | 6091476 | ||
KJ534919.1 | - | 4 | 491 | |||
KJ535024.1 | - | 3 | 324 | |||
KJ535025.1 | - | 4 | 491 |