Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adh7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009626.4 | NP_033756.2 | 6 | 901 | ||
GenBank mRNA | AF331802.1 | - | 6 | 899 | ||
AK009588.2 | - | 6 | 868 | |||
AK009600.1 | - | 6 | 914 | |||
AK144795.1 | - | 852 | ||||
BC047267.1 | - | 6 | 830 | 6310424 |