Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Parp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009632.2 | NP_033762.1 | 7 | 517 | ||
GenBank mRNA | AF072521.1 | - | 7 | 492 | ||
AJ007780.1 | - | 7 | 530 | |||
AK141876.1 | - | 5 | 1692 | |||
BC052000.1 | - | 6 | 902 | |||
BC062150.1 | - | 7 | 517 | 6333875 |