Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aebp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF053943.1 | - | 17 | 2345 | ||
AK004712.1 | - | 13 | 1354 | |||
AK128980.1 | - | 17 | 2421 | |||
AK158276.1 | - | 17 | 2372 | |||
AK159330.1 | - | 17 | 2352 | |||
AK159342.1 | - | 17 | 2371 | |||
AK159377.1 | - | 17 | 2342 | |||
AK159409.1 | - | 17 | 2372 | |||
AK159774.1 | - | 17 | 2343 | |||
AK159957.1 | - | 17 | 2343 | |||
AK189735.1 | - | 2 | 232 | |||
AK195891.1 | - | 2 | 232 | |||
AK195983.1 | - | 2 | 232 | |||
AK212842.1 | - | 2 | 232 | |||
BC082577.1 | - | 17 | 2357 | 30648530 | ||
X80478.1 | - | 8 | 1075 |