Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHMT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166356.1 | NP_001159828.1 | 6, 7 | 901 | ||
NM_001166357.1 | NP_001159829.1 | 6, 7 | 993 | |||
NM_001166358.1 | NP_001159830.1 | 6, 7 | 892 | |||
NM_001166359.1 | NP_001159831.1 | 6, 7 | 785 | |||
NM_005412.5 | NP_005403.2 | 6, 7 | 931 | |||
NR_029415.1 | 5, 6 | 733 | ||||
NR_029416.1 | 5, 6 | 812 | ||||
NR_029417.1 | 5, 6 | 851 | ||||
NR_048562.1 | 5, 6 | 941 | ||||
XM_011538675.2 | XP_011536977.1 | 6, 7 | 871 | |||
XM_011538676.2 | XP_011536978.1 | 6, 7 | 862 | |||
XM_011538677.2 | XP_011536979.1 | 5, 6 | 772 | |||
XM_011538678.2 | XP_011536980.1 | 5, 6 | 754 | |||
GenBank mRNA | AK055053.1 | - | 3, 4 | 1581 | ||
AK223555.1 | - | 6, 7 | 793 | |||
AK295941.1 | - | 5, 6 | 682 | |||
AK296183.1 | - | 6, 7 | 745 | |||
AK297173.1 | - | 5, 6 | 693 | |||
AK298231.1 | - | 5, 6 | 586 | |||
AK301339.1 | - | 6, 7 | 658 | |||
AK301443.1 | - | 5, 6 | 755 | |||
AK303827.1 | - | 5, 6 | 607 | |||
AK315916.1 | - | 6, 7 | 985 | |||
BC011911.2 | - | 6, 7 | 735 | 4139001 | ||
BC013677.2 | - | 6, 7 | 795 | 3863368 | ||
BC025355.1 | - | 3, 4 | 318 | |||
BC032584.1 | - | 6, 7 | 774 | 5499481 | ||
BC044211.1 | - | 6, 7 | 771 | 5521082 | ||
BC091501.1 | - | 5, 6 | 655 | |||
BT006866.1 | - | 6, 7 | 725 | |||
BX647711.1 | - | 6, 7 | 852 | |||
BX648847.1 | - | 4, 5 | 333 | |||
EU668344.1 | - | 6, 7 | 735 | |||
KU178333.1 | - | 6, 7 | 725 | |||
KU178334.1 | - | 5, 6 | 361 | |||
L11932.1 | - | 5, 6 | 636 |