Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DYRK1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001396.3 | NP_001387.2 | 11 | 3734 | ||
NM_101395.2 | NP_567824.1 | 13 | 4039 | |||
NM_130436.2 | NP_569120.1 | 11 | 5102 | |||
NM_130438.2 | NP_569122.1 | 10 | 3610 | |||
XM_005260933.4 | XP_005260990.1 | 7 | 3061 | |||
XM_006723976.3 | XP_006724039.1 | 12 | 3806 | |||
XM_006723977.3 | XP_006724040.1 | 12 | 4131 | |||
XM_006723978.3 | XP_006724041.1 | 12 | 4860 | |||
XM_006723979.3 | XP_006724042.1 | 12 | 3966 | |||
XM_011529482.2 | XP_011527784.1 | 11 | 3828 | |||
XM_011529483.2 | XP_011527785.1 | 12 | 6674 | |||
XM_011529484.2 | XP_011527786.1 | 11 | 3801 | |||
XM_011529485.2 | XP_011527787.1 | 11 | 4317 | |||
XM_017028284.1 | XP_016883773.1 | 12 | 3765 | |||
XM_017028285.1 | XP_016883774.1 | 12 | 4011 | |||
XM_017028286.1 | XP_016883775.1 | 14 | 4008 | |||
GenBank mRNA | AF108830.1 | - | 13 | 4037 | ||
BC045802.1 | - | 1 | 644 | |||
D86550.1 | - | 11 | 5105 |