Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other AIRE SIRNA Assays ›
Gene Symbol:
Aire
Gene Name:
autoimmune regulator (autoimmune polyendocrinopathy candidiasis ectodermal dystrophy)
Gene Aliases:
-
Chromosome Location:
Chr.10: 78030022 - 78043610 on Build GRCm38
UniGene ID:
Mm.35300
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
162409
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Aire
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001271549.1
NP_001258478.1
2, 3
392
NM_001271550.1
NP_001258479.1
2, 3
392
NM_001271551.1
NP_001258480.1
2, 3
392
NM_001271552.1
NP_001258481.1
2, 3
392
NM_001271553.1
NP_001258482.1
2, 3
392
NM_001271554.1
NP_001258483.1
2, 3
392
NM_001271555.1
NP_001258484.1
2, 3
392
NM_001271556.1
NP_001258485.1
2, 3
392
NM_001271557.1
NP_001258486.1
2, 3
392
NM_001271558.1
NP_001258487.1
2, 3
392
NM_001271559.1
NP_001258488.1
2, 3
392
NM_009646.2
NP_033776.1
2, 3
392
GenBank mRNA
AF079536.1
-
2, 3
362
AF128115.1
-
2, 3
362
AF128116.1
-
2, 3
362
AF128117.1
-
2, 3
362
AF128118.1
-
2, 3
362
AF128119.1
-
2, 3
362
AF128120.1
-
2, 3
362
AF128121.1
-
2, 3
362
AF128122.1
-
2, 3
362
AF128123.1
-
2, 3
362
AF128124.1
-
2, 3
362
AF128125.1
-
2, 3
362
AF128772.1
-
2, 3
305
AF128773.1
-
2, 3
305
AJ132243.1
-
2, 3
305
AJ243821.1
-
2, 3
305
AK153690.1
-
3, 4
446
BC103510.1
-
2, 3
354
40039055
BC103511.1
-
2, 3
354
40039054
BC103518.1
-
2, 3
354
40039058
EU625343.3
-
2, 3
322
JF267399.1
-
2, 3
322
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
humoral immune response
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II regulatory region sequence-specific DNA binding
DNA binding
chromatin binding
zinc ion binding
histone binding
identical protein binding
transcription regulatory region DNA binding
translation regulator activity
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0