Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK035145.1 | - | 7, 8 | 915 | ||
AK042326.1 | - | 7, 8 | 919 | |||
AK043506.1 | - | 7, 8 | 909 | |||
AK078626.1 | - | 7, 8 | 915 | |||
AK079602.1 | - | 7, 8 | 890 | |||
AK128913.1 | - | 7, 8 | 852 | |||
AK146286.1 | - | 7, 8 | 908 | |||
AK150233.1 | - | 7, 8 | 907 | |||
AK150250.1 | - | 7, 8 | 920 | |||
AK150920.1 | - | 7, 8 | 907 | |||
AK150986.1 | - | 7, 8 | 889 | |||
AK151763.1 | - | 7, 8 | 907 | |||
AK151826.1 | - | 7, 8 | 890 | |||
AK153128.1 | - | 7, 8 | 889 | |||
AK160060.1 | - | 7, 8 | 909 | |||
AK163452.1 | - | 915 | ||||
AK165032.1 | - | 7, 8 | 1153 | |||
AK196319.1 | - | 1, 2 | 69 | |||
AK214889.1 | - | 1, 2 | 47 | |||
BC005476.1 | - | 7, 8 | 897 | 3600875 | ||
S71509.1 | - | 7, 8 | 868 | |||
U07235.1 | - | 7, 8 | 914 |