Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK035145.1 | - | 11 | 1486 | ||
AK042326.1 | - | 11 | 1490 | |||
AK043506.1 | - | 11 | 1480 | |||
AK078626.1 | - | 11 | 1486 | |||
AK079602.1 | - | 11 | 1461 | |||
AK128913.1 | - | 11 | 1423 | |||
AK146286.1 | - | 11 | 1479 | |||
AK150233.1 | - | 11 | 1478 | |||
AK150250.1 | - | 11 | 1489 | |||
AK150920.1 | - | 11 | 1478 | |||
AK150986.1 | - | 11 | 1460 | |||
AK151763.1 | - | 11 | 1478 | |||
AK151826.1 | - | 11 | 1463 | |||
AK153128.1 | - | 11 | 1460 | |||
AK160060.1 | - | 11 | 1480 | |||
AK163452.1 | - | 1489 | ||||
AK165032.1 | - | 11 | 1724 | |||
AK188389.1 | - | 1 | 69 | |||
AK189526.1 | - | 1 | 37 | |||
AK199304.1 | - | 1 | 37 | |||
AK206041.1 | - | 2 | 210 | |||
AK213635.1 | - | 1 | 37 | |||
BC005476.1 | - | 11 | 1468 | 3600875 | ||
S71509.1 | - | 11 | 1439 | |||
U07235.1 | - | 11 | 1485 |