Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capza1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009797.2 | NP_033927.2 | 4, 5 | 351 | ||
GenBank mRNA | AK146296.1 | - | 4, 5 | 237 | ||
AK147049.1 | - | 4, 5 | 240 | |||
AK149602.1 | - | 4, 5 | 237 | |||
AK151253.1 | - | 4, 5 | 248 | |||
AK152364.1 | - | 4, 5 | 236 | |||
AK161370.1 | - | 4, 5 | 353 | |||
AK163418.1 | - | 4, 5 | 237 | |||
AK165655.1 | - | 4, 5 | 239 | |||
AK166964.1 | - | 4, 5 | 240 | |||
AK167193.1 | - | 4, 5 | 238 | |||
AK168099.1 | - | 4, 5 | 237 | |||
AK168916.1 | - | 4, 5 | 239 | |||
AK169806.1 | - | 4, 5 | 237 | |||
BC016232.1 | - | 4, 5 | 240 | 4489604 | ||
BC085506.1 | - | 4, 5 | 226 | 6817308 | ||
U16740.1 | - | 4, 5 | 220 |