Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011121.3 | NP_035251.3 | 10 | 2124 | ||
GenBank mRNA | AK134045.1 | - | 10 | 2065 | ||
AK148050.1 | - | 10 | 2038 | |||
AK164027.1 | - | 10 | 2094 | |||
AK205623.1 | - | 1 | 224 | |||
BC006880.1 | - | 10 | 2076 | 3596381 | ||
L19558.1 | - | 10 | 2132 | |||
U01063.1 | - | 10 | 2045 |