Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011121.3 | NP_035251.3 | 9 | 1700 | ||
GenBank mRNA | AK134045.1 | - | 9 | 1641 | ||
AK148050.1 | - | 9 | 1614 | |||
AK164027.1 | - | 9 | 1670 | |||
BC006880.1 | - | 9 | 1652 | 3596381 | ||
L19558.1 | - | 9 | 1707 | |||
U01063.1 | - | 9 | 1621 |