Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfrc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011638.4 | NP_035768.1 | 5 | 621 | ||
GenBank mRNA | AK048228.1 | - | 5 | 535 | ||
AK088961.1 | - | 5 | 535 | |||
BC054522.1 | - | 5 | 506 | 6406097 | ||
X57349.1 | - | 4 | 457 |