Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Farsb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278075.1 | NP_001265004.1 | 4 | 340 | ||
NM_011811.4 | NP_035941.2 | 4 | 340 | |||
GenBank mRNA | AF123263.1 | - | 4 | 340 | ||
AK010271.1 | - | 4 | 303 | |||
AK012154.1 | - | 4 | 303 | |||
AK045432.1 | - | 4 | 292 | |||
AK076607.1 | - | 4 | 302 | |||
AK134799.1 | - | 3 | 246 | |||
AK165645.1 | - | 4 | 207 | |||
AK167945.1 | - | 4 | 303 | |||
AK168923.1 | - | 4 | 284 | |||
AK169621.1 | - | 3 | 247 | |||
BC002313.1 | - | 4 | 199 | 3592805 | ||
BC016428.1 | - | 4 | 307 | 4239906 | ||
CN693684.1 | - | 1 | 28 |