Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Creb3l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011957.2 | NP_036087.2 | 7, 8 | 1036 | ||
GenBank mRNA | AB017614.1 | - | 7, 8 | 998 | ||
AK142368.1 | - | 1037 | ||||
BC016447.1 | - | 7, 8 | 1011 | 4921584 |