Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170406.1 | NP_001163877.1 | 4 | 386 | ||
NM_001170407.1 | NP_001163878.1 | 4 | 374 | |||
NM_001320918.1 | NP_001307847.1 | 4 | 386 | |||
NM_001786.4 | NP_001777.1 | 4 | 374 | |||
NM_033379.4 | NP_203698.1 | 4 | 374 | |||
XM_005270303.3 | XP_005270360.1 | 4 | 456 | |||
GenBank mRNA | AK291939.1 | - | 4 | 311 | ||
AK295741.1 | - | 4 | 352 | |||
AW976143.1 | - | 4 | 289 | |||
BC014563.1 | - | 3 | 245 | 2967728 | ||
BC107750.1 | - | 4 | 335 | 6710672 | ||
BF685285.1 | - | 4 | 435 | |||
BG723076.1 | - | 4 | 379 | |||
BT007004.1 | - | 3 | 232 | |||
CR933728.1 | - | 4 | 333 | |||
D88357.1 | - | 3 | 259 | |||
DA823877.1 | - | 4 | 387 | |||
X05360.1 | - | 4 | 358 | |||
Y00272.1 | - | 4 | 358 |