Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abca1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013454.3 | NP_038482.3 | 12 | 1807 | ||
GenBank mRNA | AK051920.1 | - | 12 | 1808 | ||
X75926.1 | - | 11 | 1570 |