Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205049.1 | NP_001191978.1 | 3 | 360 | ||
NM_001277245.1 | NP_001264174.1 | 3 | 423 | |||
NM_001277246.1 | NP_001264175.1 | 3 | 356 | |||
NM_001277247.1 | NP_001264176.1 | 3 | 234 | |||
NM_013455.3 | NP_038483.1 | 3 | 360 | |||
GenBank mRNA | AK006695.1 | - | 3 | 360 | ||
AK160427.1 | - | 3 | 307 | |||
BC103577.1 | - | 3 | 323 | |||
BC103578.1 | - | 3 | 323 | |||
BC103579.1 | - | 3 | 386 | 40045630 | ||
BC103580.1 | - | 3 | 323 | 40045632 | ||
D00754.1 | - | 3 | 320 | |||
M85170.1 | - | 2 | 323 | |||
X52466.1 | - | 2 | 323 |