Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005439.2 | NP_005430.1 | 6 | 428 | ||
NR_026581.1 | 6 | 925 | ||||
GenBank mRNA | AF070539.1 | - | 6 | 558 | ||
AK289869.1 | - | 6 | 428 | |||
AK313998.1 | - | 6 | 729 | |||
BC000898.2 | - | 6 | 498 | 2900086 | ||
BC002340.2 | - | 6 | 538 | 2820309 | ||
BT019393.1 | - | 5 | 365 | |||
BT019432.1 | - | 5 | 365 | |||
CR541700.1 | - | 5 | 365 | |||
U57342.1 | - | 6 | 544 |