Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Add1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024458.3 | NP_001019629.2 | 6 | 825 | ||
NM_001102444.1 | NP_001095914.1 | 6 | 825 | |||
NM_013457.3 | NP_038485.1 | 6 | 825 | |||
GenBank mRNA | AF096839.1 | - | 6 | 723 | ||
AF189771.1 | - | 5 | 605 | |||
AK054558.1 | - | 6 | 677 | |||
AK079960.1 | - | 6 | 680 | |||
AK190114.1 | - | 3 | 170 | |||
AK192842.1 | - | 3 | 170 | |||
AK195469.1 | - | 3 | 170 | |||
AK207989.1 | - | 3 | 170 | |||
AK216781.1 | - | 3 | 170 | |||
BC034368.1 | - | 5 | 656 | 4008924 |