Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ahr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013464.4 | NP_038492.1 | 5 | 838 | ||
GenBank mRNA | AF405560.1 | - | 5 | 472 | ||
AF405561.1 | - | 5 | 472 | |||
AF405562.1 | - | 5 | 472 | |||
AF405563.1 | - | 5 | 472 | |||
AF405565.1 | - | 5 | 472 | |||
AF405566.1 | - | 5 | 472 | |||
AF405567.1 | - | 5 | 472 | |||
AF405568.1 | - | 5 | 472 | |||
AF405570.1 | - | 5 | 472 | |||
AK153678.1 | - | 5 | 839 | |||
AK155380.1 | - | 5 | 890 | |||
AY662080.1 | - | 5 | 472 | |||
AY662081.1 | - | 4 | 368 | |||
AY662082.1 | - | 5 | 472 | |||
AY662083.1 | - | 5 | 472 | |||
AY662084.1 | - | 5 | 472 | |||
D38416.1 | - | 5 | 487 |